Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001003653.1 | NP_001003653.1 | 3 | 892 | |
NM_001003654.1 | NP_001003654.1 | 3 | 924 | ||
XM_006255490.2 | XP_006255552.1 | 3 | 943 | ||
GenBank mRNA | AY682586.1 | - | 3 | 924 | |
AY682587.1 | - | 3 | 892 | ||
DQ011518.1 | - | 3 | 860 |