Gene Transcripts

Gene Symbol
Ly6g6f
Entrez Gene ID
309609
Gene Name
lymphocyte antigen 6 complex, locus G6F
Gene Aliases
G6f
Chromosome Location
Chr. 20: 5071047 - 5076627 on Build Rnor_6.0
UniGene ID
Rn.145028
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003691.1 NP_001003691.1 2 354
XM_006256066.2 XP_006256128.1 2 409

View Full Product Details