Gene Transcripts

Gene Symbol
Rpap3
Entrez Gene ID
300189
Gene Name
RNA polymerase II associated protein 3
Gene Aliases
Chromosome Location
Chr. 7: 139167302 - 139196355 on Build Rnor_6.0
UniGene ID
Rn.49051
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004243.1 NP_001004243.1 10 1190
XM_008765690.1 XP_008763912.1 11 1534
GenBank mRNA BC079414.1 - 10 1190

View Full Product Details