Gene Transcripts

Gene Symbol
Esam
Entrez Gene ID
300519
Gene Name
endothelial cell adhesion molecule
Gene Aliases
Chromosome Location
Chr. 8: 39997875 - 40008802 on Build Rnor_6.0
UniGene ID
Rn.17089
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004245.1 NP_001004245.1 2, 3 350
GenBank mRNA BC079093.1 - 2, 3 350

View Full Product Details