Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005543.1 | NP_001005543.1 | 7 | 639 | |
XM_006238646.2 | XP_006238708.1 | 7 | 585 | ||
XM_008763993.1 | XP_008762215.1 | 7 | 662 | ||
GenBank mRNA | BC081913.1 | - | 7 | 639 | |
FQ212041.1 | - | 6 | 579 |