Gene Transcripts
- Gene Symbol
- DAAM1
- Entrez Gene ID
- 23002
- Gene Name
- dishevelled associated activator of morphogenesis 1
- Gene Aliases
- Chromosome Location
- Chr. 14: 59188611 - 59371405 on Build GRCh38
- UniGene ID
- Hs.19156
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270520.1 |
NP_001257449.1
|
3, 4 |
424 |
|
NM_014992.2 |
NP_055807.1
|
2, 3 |
318 |
|
XM_005267430.1 |
XP_005267487.1
|
3, 4 |
476 |
|
XM_005267431.1 |
XP_005267488.1
|
3, 4 |
576 |
|
GenBank mRNA |
AB014566.1 |
- |
2, 3 |
304 |
|
AK291790.1 |
- |
3, 4 |
393 |
|
BC038428.1 |
- |
3, 4 |
416 |
4993504 |
BC064999.1 |
- |
3, 4 |
407 |
5784628 |
BP199650.1 |
- |
3, 4 |
424 |
|
BQ050640.1 |
- |
3, 4 |
406 |
|
BX247986.1 |
- |
3, 4 |
501 |
|
View Full Product Details