Gene Transcripts

Gene Symbol
TRIM2
Entrez Gene ID
23321
Gene Name
tripartite motif containing 2
Gene Aliases
CMT2R, RNF86
Chromosome Location
Chr. 4: 153152281 - 153339322 on Build GRCh38
UniGene ID
Hs.435711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130067.1 NP_001123539.1 12 2706
NM_015271.4 NP_056086.2 12 2703
XM_006714157.1 XP_006714220.1 12 2715
XM_006714158.1 XP_006714221.1 12 2680
XM_006714159.3 XP_006714222.1 12 3526
XM_006714160.1 XP_006714223.1 13 3480
XM_006714161.1 XP_006714224.1 13 3477
XM_006714162.1 XP_006714225.1 13 3197
XM_006714165.3 XP_006714228.1 10 2908
XM_011531794.1 XP_011530096.1 12 2794
XM_011531795.1 XP_011530097.1 13 3200
XM_011531796.2 XP_011530098.1 12 2730
XM_011531798.2 XP_011530100.1 12 2835
XM_017007943.1 XP_016863432.1 12 2668
XM_017007944.1 XP_016863433.1 13 3468
XM_017007945.1 XP_016863434.1 13 3465
XM_017007946.1 XP_016863435.1 12 3511
XM_017007947.1 XP_016863436.1 12 2782
XM_017007948.1 XP_016863437.1 12 2736
XM_017007949.1 XP_016863438.1 13 3185
XM_017007950.1 XP_016863439.1 13 3188
XM_017007951.1 XP_016863440.1 12 2882
XM_017007952.1 XP_016863441.1 12 2890
XM_017007953.1 XP_016863442.1 10 2246
XM_017007954.1 XP_016863443.1 13 2760
XM_017007955.1 XP_016863444.1 13 2753
GenBank mRNA AB011089.1 - 12 2651
AF220018.1 - 12 2651
AL110234.1 - 5 963
BC011052.1 - 12 2706 4156234

View Full Product Details