Gene Transcripts

Gene Symbol
UTP11
Entrez Gene ID
51118
Gene Name
UTP11, small subunit processome component homolog (S. cerevisiae)
Gene Aliases
CGI-94, CGI94, UTP11L
Chromosome Location
Chr. 1: 38012712 - 38024825 on Build GRCh38
UniGene ID
Hs.472038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016037.3 NP_057121.2 5, 6 531
XM_011541555.2 XP_011539857.1 4, 5 465
GenBank mRNA AF067802.1 - 5, 6 494
AF151852.1 - 5, 6 509
AK023587.1 - 3, 4 1400
AK291900.1 - 5, 6 461
AK308377.1 - 4, 5 466
BC005182.1 - 5, 6 476 3995984

View Full Product Details