Gene Transcripts

Gene Symbol
YPEL5
Entrez Gene ID
51646
Gene Name
yippee like 5
Gene Aliases
CGI-127
Chromosome Location
Chr. 2: 30146568 - 30160533 on Build GRCh38
UniGene ID
Hs.515890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127399.1 NP_001120871.1 4 1706
NM_001127400.1 NP_001120872.1 4 1942
NM_001127401.1 NP_001120873.1 5 2003
NM_016061.2 NP_057145.1 3 1645
XM_017004318.1 XP_016859807.1 5 2241
XM_017004319.1 XP_016859808.1 4 1679
XM_017004320.1 XP_016859809.1 4 2180
XM_017004321.1 XP_016859810.1 4 1872
XM_017004322.1 XP_016859811.1 4 2103
GenBank mRNA AF151885.1 - 3 1567
AK025730.1 - 4 1583

View Full Product Details