Gene Transcripts

Gene Symbol
Dpcd
Entrez Gene ID
294004
Gene Name
deleted in primary ciliary dyskinesia
Gene Aliases
RGD1307648
Chromosome Location
Chr. 1: 265298880 - 265318521 on Build Rnor_6.0
UniGene ID
Rn.50057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013905.1 NP_001013927.1 6 541
XM_006231450.2 XP_006231512.1 6 582
GenBank mRNA BC078988.1 - 6 541

View Full Product Details