Gene Transcripts

Gene Symbol
Ggnbp1
Entrez Gene ID
70772
Gene Name
gametogenetin binding protein 1
Gene Aliases
0610031G08Rik, mdf
Chromosome Location
Chr. 17: 27018035 - 27036378 on Build GRCm38
UniGene ID
Mm.440437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027544.2 NP_081820.2 1 291
GenBank mRNA AB250379.1 - 1 174
AK077129.1 - 1 174
AY141192.1 - 1 291
BC049764.1 - 1 152 6774330

View Full Product Details