Gene Transcripts

Gene Symbol
CCDC85A
Entrez Gene ID
114800
Gene Name
coiled-coil domain containing 85A
Gene Aliases
Chromosome Location
Chr. 2: 56184123 - 56386174 on Build GRCh38
UniGene ID
Hs.117136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080433.1 NP_001073902.1 6 2758
XM_005264125.1 XP_005264182.1 5 2148
GenBank mRNA AB067499.2 - 6 2845
BC131557.1 - 6 1430

View Full Product Details