Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304530.1 | NP_001291459.1 | 6 | 561 | |
NM_001304532.1 | NP_001291461.1 | 6 | 561 | ||
NM_018246.2 | NP_060716.2 | 7 | 613 | ||
NR_130761.1 | 6 | 509 | |||
NR_130762.1 | 5 | 457 | |||
XM_011544570.2 | XP_011542872.1 | 6 | 560 | ||
XM_011544571.2 | XP_011542873.1 | 5 | 439 | ||
GenBank mRNA | AK001715.1 | - | 6 | 561 | |
AK027900.1 | - | 7 | 565 | ||
AK292875.1 | - | 6 | 548 | ||
AK294848.1 | - | 6 | 496 | ||
AK311437.1 | - | 5 | 420 | ||
AK316015.1 | - | 6 | 463 | ||
BC032588.1 | - | 6 | 550 | 5502227 | |
BC049197.1 | - | 7 | 567 | 5534448 | |
BC054039.1 | - | 7 | 553 | ||
BC065295.1 | - | 7 | 554 | 6139413 | |
BC110391.1 | - | 5 | 398 | ||
BG718771.1 | - | 6 | 553 |