Gene Transcripts

Gene Symbol
C5orf22
Entrez Gene ID
55322
Gene Name
chromosome 5 open reading frame 22
Gene Aliases
Chromosome Location
Chr. 5: 31532086 - 31555058 on Build GRCh38
UniGene ID
Hs.519246
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018356.2 NP_060826.2 9 2421
NR_134298.1 9 2197
XM_006714479.2 XP_006714542.1 9 2303
XM_006714480.3 XP_006714543.1 9 2090
XM_011514062.2 XP_011512364.1 10 2499
XM_017009607.1 XP_016865096.1 11 2701
XM_017009608.1 XP_016865097.1 10 2619
XM_017009609.1 XP_016865098.1 10 2501
XM_017009610.1 XP_016865099.1 10 2387
XM_017009611.1 XP_016865100.1 10 2384
XM_017009612.1 XP_016865101.1 10 2296
XM_017009613.1 XP_016865102.1 9 2380
XM_017009614.1 XP_016865103.1 10 2275
XM_017009615.1 XP_016865104.1 10 2183
XM_017009616.1 XP_016865105.1 9 2094
GenBank mRNA AF038176.1 - 1 225
AK002055.1 - 9 2407
AL834427.1 - 4 1476
AV710784.1 - 1 108
BC032845.1 - 9 2180

View Full Product Details