Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001093728.2 | NP_001087197.1 | 9 | 1377 | |
NM_001261817.1 | NP_001248746.1 | 9 | 1362 | ||
NM_001261818.1 | NP_001248747.1 | 8 | 1239 | ||
NM_001261819.1 | NP_001248748.1 | 8 | 1224 | ||
NM_001261820.1 | NP_001248749.1 | 10 | 1583 | ||
NM_001261821.1 | NP_001248750.1 | 10 | 1568 | ||
NM_018679.5 | NP_061149.1 | 9 | 1471 | ||
XM_005249339.2 | XP_005249396.1 | 8 | 1132 | ||
XM_011514830.1 | XP_011513132.1 | 9 | 1609 | ||
XM_011514831.1 | XP_011513133.1 | 7 | 1045 | ||
XM_011514832.1 | XP_011513134.1 | 6 | 909 | ||
GenBank mRNA | AF260330.1 | - | 6 | 920 | |
AF269223.1 | - | 8 | 1155 | ||
AF536532.1 | - | 10 | 1423 | ||
AF536533.1 | - | 9 | 1410 | ||
AK057234.1 | - | 9 | 1194 | ||
AK098590.1 | - | 7 | 966 | ||
AK301778.1 | - | 9 | 1253 | ||
AK301975.1 | - | 8 | 1160 | ||
AK302022.1 | - | 8 | 1125 | ||
AK302375.1 | - | 9 | 1302 | ||
AK302456.1 | - | 9 | 1283 | ||
AK315078.1 | - | 9 | 1404 | ||
AK316403.1 | - | 9 | 1186 | ||
AY069943.1 | - | 8 | 1179 | ||
BC033729.1 | - | 9 | 1323 | 5167179 | |
BC040003.1 | - | 10 | 1425 | 5741923 | |
BC048418.1 | - | 9 | 1401 | 5298037 | |
BC090050.1 | - | 10 | 1429 | 30387768 |