Gene Transcripts

Gene Symbol
LINC-ROR
Entrez Gene ID
100885779
Gene Name
long intergenic non-protein coding RNA, regulator of reprogramming
Gene Aliases
ROR, lincRNA-RoR
Chromosome Location
Chr. 18: 57054571 - 57072119 on Build GRCh38
UniGene ID
Hs.742426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_048536.1 1, 2 210
GenBank mRNA AB908956.1 - 1, 2 210
HQ315778.1 - 1, 2 210
LC027886.1 - 1, 2 150
LC027887.1 - 1, 2 150

View Full Product Details