Gene Transcripts

Gene Symbol
ANKRD62
Entrez Gene ID
342850
Gene Name
ankyrin repeat domain 62
Gene Aliases
DKFZp779B1634
Chromosome Location
Chr. 18: 12093796 - 12181684 on Build GRCh38
UniGene ID
Hs.448906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277333.1 NP_001264262.1 5 843
XM_017025720.1 XP_016881209.1 5 896
GenBank mRNA BX648696.1 - 5 844

View Full Product Details