Gene Transcripts

Gene Symbol
KRTAP10-8
Entrez Gene ID
386681
Gene Name
keratin associated protein 10-8
Gene Aliases
KAP10.8, KRTAP18-8, KRTAP18.8
Chromosome Location
Chr. 21: 44612079 - 44612954 on Build GRCh38
UniGene ID
Hs.528021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198695.2 NP_941968.2 1 156
GenBank mRNA AB076355.1 - 1 156
BC127016.1 - 1 150 40119338

View Full Product Details