Gene Transcripts

Gene Symbol
Tex21
Entrez Gene ID
299152
Gene Name
testis expressed 21
Gene Aliases
Chromosome Location
Chr. 6: 99236653 - 99274343 on Build Rnor_6.0
UniGene ID
Rn.127173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025658.1 NP_001020829.1 4 892
XM_006240226.2 XP_006240288.1 6 1095
XM_006240227.2 XP_006240289.1 6 1139
XM_008764743.1 XP_008762965.1 6 1133
XM_008764744.1 XP_008762966.1 5 1010
XM_008764745.1 XP_008762967.1 4 615
GenBank mRNA BC097422.1 - 4 892

View Full Product Details