Gene Transcripts

Gene Symbol
Lrrc6
Entrez Gene ID
299920
Gene Name
leucine rich repeat containing 6
Gene Aliases
Lrtp, Tslrp
Chromosome Location
Chr. 7: 107130402 - 107231507 on Build Rnor_6.0
UniGene ID
Rn.124772
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025659.2 NP_001020830.2 9, 10 1236
GenBank mRNA BC097394.1 - 9, 10 1237

View Full Product Details