Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037496.1 | NP_001032585.1 | 10 | 2549 | |
XM_006256737.2 | XP_006256799.1 | 10 | 3202 | ||
XM_006256738.2 | XP_006256800.1 | 10 | 3198 | ||
XM_006256739.2 | XP_006256801.1 | 10 | 3200 | ||
XM_006256740.2 | XP_006256802.1 | 11 | 3072 | ||
XM_006256741.2 | XP_006256803.1 | 11 | 3068 | ||
GenBank mRNA | BC110054.1 | - | 10 | 2549 |