Gene Transcripts

Gene Symbol
Defb49
Entrez Gene ID
641653
Gene Name
defensin beta 49
Gene Aliases
Chromosome Location
Chr. 9: 24446265 - 24451435 on Build Rnor_6.0
UniGene ID
Rn.156588
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037527.1 NP_001032616.1 1, 2 50
GenBank mRNA AY621372.1 - 1, 2 50

View Full Product Details