Gene Transcripts

Gene Symbol
Spag11b
Entrez Gene ID
652998
Gene Name
sperm associated antigen 11b
Gene Aliases
Spag11c
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.155415
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037850.1 NP_001032939.1 2, 3 218
NM_001037852.1 NP_001032941.1 2, 3 278
GenBank mRNA DQ012092.1 - 2, 3 218
DQ012093.1 - 2, 3 278

Gene Symbol
Spag11bl
Entrez Gene ID
100362580
Gene Name
sperm associated antigen 11b-like
Gene Aliases
Chromosome Location
Chr. 16: 75145234 - 75148001 on Build Rnor_6.0
UniGene ID
Rn.155415
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728419.4 XP_002728465.2 2, 3 405
XM_006253377.2 XP_006253439.1 2, 3 408
GenBank mRNA AY600145.1 - 2, 3 238

View Full Product Details