Gene Transcripts
- Gene Symbol
- KLHL13
- Entrez Gene ID
- 90293
- Gene Name
- kelch like family member 13
- Gene Aliases
- BKLHD2
- Chromosome Location
- Chr. X: 117897813 - 118117340 on Build GRCh38
- UniGene ID
- Hs.348262
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168299.1 |
NP_001161771.1
|
4, 5 |
510 |
|
NM_001168300.1 |
NP_001161772.1
|
3, 4 |
758 |
|
NM_001168301.1 |
NP_001161773.1
|
4, 5 |
408 |
|
NM_001168302.1 |
NP_001161774.1
|
4, 5 |
702 |
|
NM_001168303.1 |
NP_001161775.1
|
3, 4 |
628 |
|
NM_033495.3 |
NP_277030.2
|
3, 4 |
1278 |
|
XM_011531409.2 |
XP_011529711.1
|
5, 6 |
1170 |
|
XM_011531410.2 |
XP_011529712.1
|
5, 6 |
470 |
|
XM_011531411.1 |
XP_011529713.1
|
4, 5 |
849 |
|
XM_017029950.1 |
XP_016885439.1
|
3, 4 |
1539 |
|
GenBank mRNA |
AB037730.1 |
- |
2, 3 |
2637 |
|
AK122724.1 |
- |
3, 4 |
345 |
|
AK125356.1 |
- |
3, 4 |
1278 |
|
AK296324.1 |
- |
4, 5 |
417 |
|
AK299257.1 |
- |
3, 4 |
522 |
|
AK302713.1 |
- |
4, 5 |
408 |
|
AK316509.1 |
- |
4, 5 |
662 |
|
AL591986.1 |
- |
3, 4 |
425 |
|
BC064576.1 |
- |
3, 4 |
430 |
5727768 |
DC354391.1 |
- |
4, 5 |
510 |
|
View Full Product Details