Gene Transcripts

Gene Symbol
UBXN10
Entrez Gene ID
127733
Gene Name
UBX domain protein 10
Gene Aliases
UBXD3
Chromosome Location
Chr. 1: 20183106 - 20196060 on Build GRCh38
UniGene ID
Hs.432503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152376.4 NP_689589.1 2 270
XM_005245742.3 XP_005245799.1 2 774
XM_011540699.2 XP_011539001.1 2 478
GenBank mRNA AK058158.1 - 2 255
BC047428.1 - 2 265 5295853
BX648631.1 - 2 260
DB061550.1 - 2 270

View Full Product Details