Gene Transcripts
- Gene Symbol
- MROH2B
- Entrez Gene ID
- 133558
- Gene Name
- maestro heat like repeat family member 2B
- Gene Aliases
- HEATR7B2
- Chromosome Location
- Chr. 5: 40998020 - 41071342 on Build GRCh38
- UniGene ID
- Hs.97714
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173489.4 |
NP_775760.3
|
28, 29 |
3382 |
|
XM_011513952.1 |
XP_011512254.1
|
28, 29 |
3368 |
|
XM_011513953.1 |
XP_011512255.1
|
27, 28 |
3306 |
|
GenBank mRNA |
AK097562.1 |
- |
4, 5 |
428 |
|
AK126005.1 |
- |
13, 14 |
1251 |
|
AK128183.1 |
- |
18, 19 |
1848 |
|
AL832763.1 |
- |
23, 24 |
2307 |
|
BC044937.1 |
- |
16, 17 |
1854 |
|
BC052962.1 |
- |
28, 29 |
3343 |
4838071 |
CR749346.1 |
- |
23, 24 |
2283 |
|
View Full Product Details