Gene Transcripts

Gene Symbol
RAB7A
Entrez Gene ID
7879
Gene Name
RAB7A, member RAS oncogene family
Gene Aliases
PRO2706, RAB7
Chromosome Location
Chr. 3: 128726136 - 128814798 on Build GRCh38
UniGene ID
Hs.684374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004637.5 NP_004628.4 6 1170
GenBank mRNA AK094449.1 - 5 851
AK225177.1 - 5 924
AK225691.1 - 5 917
AK290721.1 - 6 1074
AK308934.1 - 6 972
BC008721.2 - 6 1113 2821435
BC013728.1 - 5 953 3882701
BX537775.1 - 4 733

View Full Product Details