Gene Transcripts
- Gene Symbol
- SFRP4
- Entrez Gene ID
- 6424
- Gene Name
- secreted frizzled related protein 4
- Gene Aliases
- FRP-4, FRPHE, sFRP-4
- Chromosome Location
- Chr. 7: 37905932 - 37916923 on Build GRCh38
- UniGene ID
- Hs.658169
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003014.3 |
NP_003005.2
|
1, 2 |
838 |
|
GenBank mRNA |
AF026692.1 |
- |
1, 2 |
709 |
|
AK302357.1 |
- |
1, 2 |
760 |
|
AK303977.1 |
- |
2, 3 |
658 |
|
AK313029.1 |
- |
1, 2 |
728 |
|
BC047684.1 |
- |
1, 2 |
696 |
5742936 |
BC058911.1 |
- |
1, 2 |
716 |
5228231 |
BT019679.1 |
- |
1, 2 |
452 |
|
CR541731.1 |
- |
1, 2 |
452 |
|
CR541755.1 |
- |
1, 2 |
452 |
|
DQ420628.1 |
- |
1, 2 |
471 |
|
View Full Product Details