Gene Transcripts

Gene Symbol
SFRP4
Entrez Gene ID
6424
Gene Name
secreted frizzled related protein 4
Gene Aliases
FRP-4, FRPHE, sFRP-4
Chromosome Location
Chr. 7: 37905932 - 37916923 on Build GRCh38
UniGene ID
Hs.658169
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003014.3 NP_003005.2 6 1957
GenBank mRNA AF026692.1 - 6 1826
BC058911.1 - 6 1835 5228231

View Full Product Details