Gene Transcripts
- Gene Symbol
- Pydc3
- Entrez Gene ID
- 100033459
- Gene Name
- pyrin domain containing 3
- Gene Aliases
- 9830148g24rik, E430029J22Rik
- Chromosome Location
- Chr. 1: 173673628 - 173698392 on Build GRCm38
- UniGene ID
- Mm.303377
- Species
- Mus musculus
- Gene Symbol
- Pydc4
- Entrez Gene ID
- 623121
- Gene Name
- pyrin domain containing 4
- Gene Aliases
- E030037K03Rik, E430029J22Rik
- Chromosome Location
- Chr. 1: 173566595 - 173599302 on Build GRCm38
- UniGene ID
- Mm.138449
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK145253.1 |
- |
5 |
1186 |
|
AK165868.1 |
- |
3 |
2582 |
|
View Full Product Details