Gene Transcripts

Gene Symbol
Krtap10-4
Entrez Gene ID
100191037
Gene Name
keratin associated protein 10-4
Gene Aliases
675238, A030009A09Rik, AA589524
Chromosome Location
Chr. 10: 77826169 - 77827070 on Build GRCm38
UniGene ID
Mm.432613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135991.1 NP_001129463.1 1 712
GenBank mRNA AK020700.1 - 1 740
BC115499.1 - 1 712 40100932
BC115500.1 - 1 712 40100933

View Full Product Details