Gene Transcripts

Gene Symbol
Neb
Entrez Gene ID
17996
Gene Name
nebulin
Gene Aliases
AI595938
Chromosome Location
Chr. 2: 52136640 - 52338798 on Build GRCm38
UniGene ID
Mm.389247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010889.1 NP_035019.1 1 229
GenBank mRNA BG086842.2 - 1 229

View Full Product Details