Gene Transcripts

Gene Symbol
Shroom4
Entrez Gene ID
208431
Gene Name
shroom family member 4
Gene Aliases
D430043L16Rik, Gm724, Shrm4
Chromosome Location
Chr. X: 6399938 - 6637452 on Build GRCm38
UniGene ID
Mm.23208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040459.1 NP_001035549.1 6 3058
GenBank mRNA AK052531.1 - 5 3101
AK173129.1 - 5 3080
BC151022.1 - 6 3349 9087935
DQ435686.1 - 6 3058
DQ435687.1 - 6 3058

View Full Product Details