Gene Transcripts

Gene Symbol
Rmi2
Entrez Gene ID
223970
Gene Name
RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae)
Gene Aliases
A630055G03Rik, Gm537, Gm929
Chromosome Location
Chr. 16: 10835059 - 10843235 on Build GRCm38
UniGene ID
Mm.325777
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162932.1 NP_001156404.1 2 861
GenBank mRNA AK143588.1 - 2 858

View Full Product Details