Gene Transcripts

Gene Symbol
Lce6a
Entrez Gene ID
78382
Gene Name
late cornified envelope 6A
Gene Aliases
2210420J11Rik
Chromosome Location
Chr. 3: 92604763 - 92621660 on Build GRCm38
UniGene ID
Mm.235474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166172.1 NP_001159644.1 3 507
GenBank mRNA AK019072.1 - 2 203

View Full Product Details