Gene Transcripts
- Gene Symbol
- Rps14
- Entrez Gene ID
- 29284
- Gene Name
- ribosomal protein S14
- Gene Aliases
- Chromosome Location
- Chr. 18: 56042532 - 56047316 on Build Rnor_6.0
- UniGene ID
- Rn.154712
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022672.1 |
NP_073163.1
|
2, 3 |
194 |
|
GenBank mRNA |
X15040.1 |
- |
2, 3 |
194 |
|
View Full Product Details