Gene Transcripts
- Gene Symbol
- ROR2
- Entrez Gene ID
- 4920
- Gene Name
- receptor tyrosine kinase like orphan receptor 2
- Gene Aliases
- BDB, BDB1, NTRKR2
- Chromosome Location
- Chr. 9: 91722596 - 91950206 on Build GRCh38
- UniGene ID
- Hs.644776
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318204.1 |
NP_001305133.1
|
2, 3 |
382 |
|
NM_004560.3 |
NP_004551.2
|
2, 3 |
382 |
|
XM_005252008.4 |
XP_005252065.1
|
2, 3 |
347 |
|
XM_006717121.3 |
XP_006717184.1
|
2, 3 |
1004 |
|
XM_017014762.1 |
XP_016870251.1
|
2, 3 |
2079 |
|
XM_017014763.1 |
XP_016870252.1
|
2, 3 |
258 |
|
GenBank mRNA |
AB209154.1 |
- |
4, 5 |
651 |
|
BC033697.1 |
- |
2, 3 |
369 |
|
BC051273.1 |
- |
2, 3 |
194 |
|
BC130522.1 |
- |
2, 3 |
270 |
40146553 |
M97639.1 |
- |
2, 3 |
382 |
|
View Full Product Details