Gene Transcripts

Gene Symbol
Rtn1
Entrez Gene ID
116644
Gene Name
reticulon 1
Gene Aliases
Nsp
Chromosome Location
Chr. 6: 94951016 - 95177477 on Build Rnor_6.0
UniGene ID
Rn.55126
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053865.1 NP_446317.1 5 2510
XM_008764690.1 XP_008762912.1 3 540
GenBank mRNA FQ212949.1 - 3 451
FQ214343.1 - 3 376
U17603.1 - 3 510
U17604.1 - 5 2510

View Full Product Details