Gene Transcripts

Gene Symbol
Pla2g4a
Entrez Gene ID
24653
Gene Name
phospholipase A2, group IVA (cytosolic, calcium-dependent)
Gene Aliases
Pla2c, Pla2g4, cPLA2
Chromosome Location
Chr. 13: 67062252 - 67206688 on Build Rnor_6.0
UniGene ID
Rn.10162
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133551.2 NP_598235.2 4, 5 428
GenBank mRNA BC070940.1 - 4, 5 428
S77829.1 - 4, 5 414
U38376.1 - 4, 5 429

View Full Product Details