Gene Transcripts

Gene Symbol
Cox6a1
Entrez Gene ID
12861
Gene Name
cytochrome c oxidase subunit VIa polypeptide 1
Gene Aliases
VIaL
Chromosome Location
Chr. 5: 115345654 - 115348955 on Build GRCm38
UniGene ID
Mm.43415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK166496.1 - 3 513
BC003807.1 - 3 511 3487598
BC052816.1 - 3 517 6439987
L06465.1 - 3 510

View Full Product Details