Gene Transcripts

Gene Symbol
Gbas
Entrez Gene ID
14467
Gene Name
glioblastoma amplified sequence
Gene Aliases
AV006093, Nipsnap2
Chromosome Location
Chr. 5: 129725044 - 129758327 on Build GRCm38
UniGene ID
Mm.12468
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008095.4 NP_032121.3 2 233
GenBank mRNA AJ001261.1 - 2 211
AK170336.1 - 2 179
BC055893.1 - 2 182 4023066
BY292750.1 - 2 234

View Full Product Details