Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290837.1 | NP_001277766.1 | 3 | 436 | |
NM_011354.3 | NP_035484.1 | 3 | 413 | ||
GenBank mRNA | AA423654.1 | - | 3 | 352 | |
AF073297.1 | - | 3 | 194 | ||
AK002351.1 | - | 3 | 189 | ||
AK008802.1 | - | 3 | 202 | ||
AK010749.1 | - | 3 | 245 | ||
AK012395.1 | - | 3 | 206 | ||
AK012603.1 | - | 3 | 192 | ||
AK018746.1 | - | 3 | 189 | ||
AK132746.1 | - | 2 | 843 | ||
AK151362.1 | - | 3 | 206 | ||
AK168858.1 | - | 3 | 193 | ||
AK169106.1 | - | 3 | 195 | ||
BC081455.1 | - | 3 | 197 | ||
BC092136.1 | - | 3 | 226 | 6814342 | |
BY070249.1 | - | 3 | 252 |