Gene Transcripts
- Gene Symbol
- Stau1
- Entrez Gene ID
- 20853
- Gene Name
- staufen (RNA binding protein) homolog 1 (Drosophila)
- Gene Aliases
- 5830401L18Rik, AW549911, C85792, Stau
- Chromosome Location
- Chr. 2: 166947549 - 166996299 on Build GRCm38
- UniGene ID
- Mm.73276
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001109905.2 |
NP_001103375.1
|
3, 4 |
572 |
|
NM_001109906.2 |
NP_001103376.1
|
3, 4 |
572 |
|
NM_001291160.1 |
NP_001278089.1
|
3, 4 |
572 |
|
NM_011490.4 |
NP_035620.1
|
3, 4 |
572 |
|
GenBank mRNA |
AF061942.1 |
- |
3, 4 |
572 |
|
AF395842.1 |
- |
2, 3 |
293 |
|
AK005002.2 |
- |
3, 4 |
513 |
|
AK139532.1 |
- |
3, 4 |
519 |
|
AK145826.1 |
- |
3, 4 |
539 |
|
AK151835.1 |
- |
3, 4 |
378 |
|
AK152269.1 |
- |
3, 4 |
384 |
|
AK152744.1 |
- |
3, 4 |
378 |
|
AK159602.1 |
- |
3, 4 |
489 |
|
AK159781.1 |
- |
3, 4 |
542 |
|
AK160979.1 |
- |
3, 4 |
513 |
|
AK167637.1 |
- |
3, 4 |
513 |
|
AK193147.1 |
- |
|
179 |
|
AY391773.1 |
- |
2, 3 |
305 |
|
BC012959.1 |
- |
3, 4 |
536 |
4188394 |
BC082277.1 |
- |
3, 4 |
509 |
|
View Full Product Details