Gene Transcripts

Gene Symbol
Stau1
Entrez Gene ID
20853
Gene Name
staufen (RNA binding protein) homolog 1 (Drosophila)
Gene Aliases
5830401L18Rik, AW549911, C85792, Stau
Chromosome Location
Chr. 2: 166947549 - 166996299 on Build GRCm38
UniGene ID
Mm.73276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109905.2 NP_001103375.1 12 2515
NM_001109906.2 NP_001103376.1 12 2533
NM_001291160.1 NP_001278089.1 12 2559
NM_011490.4 NP_035620.1 12 2509
GenBank mRNA AF061942.1 - 12 2509
AK005002.2 - 12 2456
AK139532.1 - 12 2462
AK145826.1 - 12 2500
AK151835.1 - 12 2365
AK152744.1 - 12 2365
AK158952.1 - 7 3096
AK159602.1 - 12 2450
AK159781.1 - 12 2485
AK160979.1 - 12 2474
AK167637.1 - 12 2456
BC012959.1 - 12 2479 4188394
BC082277.1 - 12 2452

View Full Product Details