Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013877.3 | NP_038905.1 | 2, 3 | 242 | |
GenBank mRNA | AF169161.1 | - | 2, 3 | 203 | |
AK044544.1 | - | 2, 3 | 243 | ||
AK087416.1 | - | 2, 3 | 103 | ||
BC018253.1 | - | 2, 3 | 265 | 4504283 |