Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163671.1 | NP_001157143.1 | 8 | 1051 | |
NM_001163672.1 | NP_001157144.1 | 8 | 1051 | ||
NM_016961.3 | NP_058657.1 | 8 | 1051 | ||
NM_207692.2 | NP_997575.2 | 8 | 1051 | ||
GenBank mRNA | AB005664.1 | - | 7 | 825 | |
AF052466.1 | - | 7 | 787 | ||
AF052467.1 | - | 7 | 787 | ||
AF052468.1 | - | 7 | 787 | ||
AF052469.1 | - | 7 | 787 | ||
AK031959.1 | - | 8 | 1050 | ||
AK031974.1 | - | 6 | 605 | ||
AK145083.1 | - | 8 | 1026 | ||
BC028341.1 | - | 8 | 1033 | 4192796 |