Gene Transcripts

Gene Symbol
Cyp11a1
Entrez Gene ID
13070
Gene Name
cytochrome P450, family 11, subfamily a, polypeptide 1
Gene Aliases
Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc
Chromosome Location
Chr. 9: 58015017 - 58027023 on Build GRCm38
UniGene ID
Mm.302865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019779.3 NP_062753.3 1 129
GenBank mRNA AF195119.1 - 1 129
AK077945.1 - 1 150
BC068264.1 - 1 148 30014890

View Full Product Details