Gene Transcripts

Gene Symbol
Ccnl1
Entrez Gene ID
56706
Gene Name
cyclin L1
Gene Aliases
2610030E23Rik, AU018493, Ccnl, ania-6a
Chromosome Location
Chr. 3: 65945912 - 65958971 on Build GRCm38
UniGene ID
Mm.175612
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019937.3 NP_064321.2 11 1846
GenBank mRNA AF159159.1 - 11 1799
BC007177.1 - 10 3839
BC069909.1 - 12 1930
BC094383.1 - 11 1782 30604933
BC117816.1 - 12 1739

View Full Product Details