Gene Transcripts

Gene Symbol
Fn3k
Entrez Gene ID
63828
Gene Name
fructosamine 3 kinase
Gene Aliases
2310074G21Rik
Chromosome Location
Chr. 11: 121434809 - 121450491 on Build GRCm38
UniGene ID
Mm.266448
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022014.4 NP_071297.1 6 728
GenBank mRNA AJ404616.1 - 6 714
AK010151.1 - 6 722
BU571242.1 - 4 357

View Full Product Details