Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025444.2 | NP_079720.1 | 4 | 244 | |
GenBank mRNA | AK007227.1 | - | 4 | 250 | |
AK007339.1 | - | 4 | 245 | ||
AK076595.1 | - | 244 | |||
AK160171.1 | - | 4 | 244 | ||
BC021447.1 | - | 4 | 256 | 5050736 |